Learn More
Biomatik Corporation Human Ubiquilin 2 (UBQLN2) ELISA Kit
Supplier:  Biomatik Corporation EKU08009
Encompass_Preferred
Human Ubiquilin 2 (UBQLN2) ELISA Kit for detecting UBQLN2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.64ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UHD9. Gene ID: 29978. Target Synonyms: CHAP1; CHAP1/DSK2; Dsk2; LIC-2; N4BP4; PLIC-2; PLIC2; NEDD4 Binding Protein 4; Protein linking IAP with cytoskeleton 2; Ubiquitin-like product Chap1/Dsk2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8705