Learn More
Biomatik Corporation Human Ubiquitin Associated Protein 2 (UBAP2) ELISA Kit
Supplier:  Biomatik Corporation EKU08014
Encompass_Preferred
Human Ubiquitin Associated Protein 2 (UBAP2) ELISA Kit for detecting UBAP2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.136ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5T6F2. Gene ID: 55833. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7351