Learn More
Biomatik Corporation Human Ubiquitin Conjugating Enzyme E2I (UBE2I) ELISA Kit
Supplier:  Biomatik Corporation EKU08019
Encompass_Preferred
Human Ubiquitin Conjugating Enzyme E2I (UBE2I) ELISA Kit for detecting UBE2I in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P63279. Target Synonyms: P18; UBC9; UBCE9; SUMO-protein ligase; Ubiquitin carrier protein 9; Ubiquitin carrier protein I; Ubiquitin-protein ligase I; SUMO-conjugating enzyme UBC9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4593