Learn More
Biomatik Corporation Human Ubiquitin Conjugating Enzyme E2C (UBE2C) ELISA Kit
Supplier:  Biomatik Corporation EKU08018
Encompass_Preferred
Human Ubiquitin Conjugating Enzyme E2C (UBE2C) ELISA Kit for detecting UBE2C in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00762. Gene ID: 11065. Target Synonyms: UBCH10; Ubiquitin carrier protein C; Ubiquitin-protein ligase C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4744