Learn More
Biomatik Corporation Human Ubiquitin Protein Ligase E3 Component N-Recognin 4 (UBR4) ELISA Kit
Supplier:  Biomatik Corporation EKU08023
Encompass_Preferred
Human Ubiquitin Protein Ligase E3 Component N-Recognin 4 (UBR4) ELISA Kit for detecting UBR4 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5T4S7. Target Synonyms: RBAF600; p600; ZUBR1; Zinc Finger,UBR1 Type 1; N-recognin-4; 600 kDa retinoblastoma protein-associated factor; Retinoblastoma-associated factor of 600 kDa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8703