Learn More
Biomatik Corporation Human Ubiquitin Specific Peptidase 1 (USP1) ELISA Kit
Supplier:  Biomatik Corporation EKU11999
Encompass_Preferred
Human Ubiquitin Specific Peptidase 1 (USP1) ELISA Kit for detecting USP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O94782. Gene ID: 7398. Target Synonyms: UBP; Ubiquitin Carboxyl-Terminal Hydrolase 1; Ubiquitin Specific Protease 1; Deubiquitinating enzyme 1; Ubiquitin thioesterase 1; Ubiquitin-specific-processing protease 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8053