Learn More
Sino Biological Human UCHL1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11663R104100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human UCHL1 (rh UCHL1; Catalog#11663-H07E; NP_004172.2; Gln 2-Ala 223).
Catalog No. 50-237-3519