Learn More
Sino Biological Anti-UCHL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11663RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human UCHL1 (rh UCHL1; Catalog#11663-H07E; NP_004172.2; Gln 2-Ala 223). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2074