Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human UCMA Synthetic Peptide

Catalog No. PEP0882 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0882 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0882 Supplier Invitrogen™ Supplier No. PEP0882
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 17 amino acids near the carboxy terminus of human UCMA. PEP-0882 can be used as a blocking peptide with polyclonal antibody PA5-20768.

UCMA is a gene that encodes for a protein that is specific to chondrocytes and highly charged. This protein is expressed abundantly in the upper immature zone of fetal and juvenile epiphyseal cartilage. The encoded protein undergoes proteolytic processing to generate a mature protein that is secreted into the extracellular matrix. The glutamic acid residues in the protein undergo gamma carboxylation in a vitamin K-dependent manner. Undercarboxylation of the protein has been associated with osteoarthritis in humans. Alternative splicing of the gene results in multiple transcript variants that encode different isoforms. UCMA is also associated with diseases such as Osteoarthritis and Poland Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 221044
Species Human
Purification Method purified
Accession Number Q8WVF2
Common Name UCMA
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 1110017I16Rik; AW121955; C10orf49; Gla-rich protein; GRP; GRP/UCMA; RGD1308977; Ucma; ucma {ECO:0000250; Ucma-C; UniProtKB:Q14BU0}; Unique cartilage matrix-associated protein; unique cartilage matrix-associated protein {ECO:0000250; Unique cartilage matrix-associated protein C-terminal fragment; upper zone of growth plate and cartilage matrix associated; upper zone of growth plate and cartilage matrix associated protein
Gene Symbol UCMA
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.