Learn More
BIOHIPPO Human ucOC Microsample
SDP
Catalog No. 5000313874
Encompass
Human ucOC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ucOC in serum plasma tissue homogenates It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 111 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ucOC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ucOC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ucOC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13874 Supplier BIOHIPPO Supplier No. BHE15210539
Add to Cart
Add to Cart
Human ucOC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ucOC in serum plasma tissue homogenates It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 111 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ucOC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ucOC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ucOC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color