Learn More
Sino Biological Anti-UCP2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102134T02100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human UCP2, and purified by antigen affinity chromatography.
Catalog No. 50-241-0028