Learn More
Biomatik Corporation Human UDP Glucose-6-Dehydrogenase (UGDH) ELISA Kit
Catalog No. 502416072
Encompass_Preferred
Human UDP Glucose-6-Dehydrogenase (UGDH) ELISA Kit for detecting UGDH in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60701. Gene ID: 7358. Target Synonyms: GDH; UDP-GlcDH; UDPGDH; UGD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6072 Supplier Biomatik Corporation Supplier No. EKU12063
Human UDP Glucose-6-Dehydrogenase (UGDH) ELISA Kit for detecting UGDH in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60701. Gene ID: 7358. Target Synonyms: GDH; UDP-GlcDH; UDPGDH; UGD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.