Learn More
Biomatik Corporation Human UDP-Glucose Glycoprotein Glucosyltransferase 1 (UGGT1) ELISA Kit
Supplier:  Biomatik Corporation EKU08034
Encompass_Preferred
Human UDP-Glucose Glycoprotein Glucosyltransferase 1 (UGGT1) ELISA Kit for detecting UGGT1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NYU2. Gene ID: 56886. Target Synonyms: UGTR; HUGT1; GT; UGCGL1; UGGT; UGT1; UDP-glucose ceramide glucosyltransferase-like 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5342