Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ULBP-1 ELISA Kit

Catalog No. EH476RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH476RBX5 5 x 96 Tests
EH476RBX10 10 x 96 Tests
EH476RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH476RBX5 Supplier Invitrogen™ Supplier No. EH476RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human ULBP-1 quantitates human ULBP-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ULBP-1.

ULBP1 is the ligand for the NKG2D receptor, together with at least ULBP2 and ULBP3. ULBPs activate multiple signaling pathways in primary NK cells, resulting in the production of cytokines and chemokines. Binding of ULBPs ligands to NKG2D induces calcium mobilization and activation of the JAK2, STAT5, ERK and PI3K kinase/Akt signal transduction pathway. In CMV infected cells, interacts with soluble CMV glycoprotein UL16. The interaction with UL16 blocked the interaction with the NKG2D receptor, providing a mechanism by which CMV infected cells might escape the immune system. UL16 also causes ULBP1 to be retained in the ER and cis-Golgi apparatus so that it does not reach the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BZM6
Assay Range 0.4-100 ng/mL
Assay Sensitivity 0.4 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias A430108B07Rik; ALCAN-beta; MULT1; N2DL1; N2DL-1; NKG2D ligand; NKG2D ligand 1; NKG2D ligand1; NKG2DL1; RAET1I; retinoic acid early transcript 1I; UL16 binding protein 1; UL16-binding protein 1; UL16-binding protein-like transcript 1; ULBP; ULBP 1; ULBP1A430108B07Rik; ALCAN-beta; MULT1; N2DL1; N2DL-1; NKG2D ligand; NKG2D ligand 1; NKG2D ligand1; NKG2DL1; RAET1I; retinoic acid early transcript 1I; UL16 binding protein 1; UL16-binding protein 1; UL16-binding protein-like transcript 1; ULBP; ULBP 1; ULBP1;
Gene ID (Entrez) 80329
Gene Symbol ULBP1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target ULBP1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.