Learn More
Sino Biological Recombinant Human ULBP1/NKG2DL1 Protein (His Tag) 1 mg
Supplier:  Sino Biological 10679H08H1
Encompass_Preferred
A DNA sequence encoding the mature form of human ULBP1 (NP_079494.1) (Met 1-Gly 216) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7200