Learn More
Sino Biological Recombinant Human ULBP1/NKG2DL1 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10679H27HB20
Encompass_Preferred
A DNA sequence encoding the human ULBP1 (NP_079494.1) (Met1-Gly216) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5222