Learn More
Sino Biological Recombinant Anti-ULBP2/NKG2DL2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 12143R02220
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ULBP2 / N2DL-2 (rh ULBP2 / N2DL-2; Catalog#12143-H08H; Q9BZM5; Met1-Ser217).
Catalog No. 50-240-8311