Learn More
Sino Biological Human ulBP2 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 12143R003100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ULBP2 (rh ULBP2; Catalog#12143-H08H; Q9BZM5; Met1-Ser217).
Catalog No. 50-237-1471