Learn More
Sino Biological Anti-ULBP2/NKG2DL2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12143RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ULBP2 (rh ULBP2; Catalog#12143-H08H; Q9BZM5; Met1-Ser217). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1194