Learn More
Sino Biological Anti-ULBP2/NKG2DL2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12143T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ULBP2 (rh ULBP2; Catalog#12143-H08H; Q9BZM5; Met1-Ser217). ULBP2 specific IgG was purified by Human ULBP2 affinity chromatography.
Catalog No. 50-240-8307