Learn More
Sigma Aldrich Fine Chemicals Biosciences Human Umbilical Vein Endot
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 200P05N
Encompass
Human Umbilical Vein Endothelial Cells (HUVEC) provide a classic model system to study many aspects of endothelial function and disease such as normal abnormal and tumor-associated angiogenesis oxidative stress hypoxia and inflammation related pathways in endothelia under normal and pathological conditions cardiovascular-related complications associated with various diseases mode of action and cardiovascular protection effects of various compounds etc.
Catalog No. 50-290-5813