Learn More
BIOHIPPO Human UNC5B Microsample
SDP
Catalog No. 5000315512
Encompass
Human UNC5B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of UNC5B in serum plasma saliva tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human UNC5B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human UNC5B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human UNC5B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15512 Supplier BIOHIPPO Supplier No. BHE15210486
Human UNC5B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of UNC5B in serum plasma saliva tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human UNC5B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human UNC5B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human UNC5B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color