Learn More
Biomatik Corporation Human Uncoupling Protein 4, Mitochondrial (UCP4) ELISA Kit
Supplier:  Biomatik Corporation EKU12403
Encompass_Preferred
Human Uncoupling Protein 4, Mitochondrial (UCP4) ELISA Kit for detecting UCP4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95847. Gene ID: 9481. Target Synonyms: SLC25A27; Solute Carrier Family 25 Member 27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5701