Learn More
ACROBiosystems Human uPAR (209-303), His Tag (UPR-H52H6) is expressed from human 293 cells (HEK293). It contains AA Leu 209 - Arg 303 (Accession # Q03405-1).
Catalog No. 502253994
Encompass_Preferred
Urokinase plasminogen activator surface receptor (U-PAR) is also known as PLAUR, Monocyte activation antigen Mo3, CD antigen CD87. PLAUR contains three UPAR/Ly6 domains. U-PAR is expressed in neurons of the rolandic area of the brain (at protein level) and is also expressed in the brain. PLAUR / CD87 interacts with MRC2, SRPX2 and SORL1. PLAUR / UPAR acts as a receptor for urokinase plasminogen activator and plays a role in localizing and promoting plasmin formation. U-PAR mediates the proteolysis-independent signal transduction activation effects of U-PA.
This item is not returnable. View return policy
Catalog No. 50-225-3994 Supplier ACROBiosystems Supplier No. UPRH52H6100UG
Urokinase plasminogen activator surface receptor (U-PAR) is also known as PLAUR, Monocyte activation antigen Mo3, CD antigen CD87. PLAUR contains three UPAR/Ly6 domains. U-PAR is expressed in neurons of the rolandic area of the brain (at protein level) and is also expressed in the brain. PLAUR / CD87 interacts with MRC2, SRPX2 and SORL1. PLAUR / UPAR acts as a receptor for urokinase plasminogen activator and plays a role in localizing and promoting plasmin formation. U-PAR mediates the proteolysis-independent signal transduction activation effects of U-PA.