Learn More
ACROBiosystems Human uPAR 25tests
Catalog No. 502534005
Encompass_Preferred
APC-Labeled Human uPAR Protein His Tag (UPR-HA2H9) is produced via conjugation of APC to Human uPAR Protein His Tag with a new generation site-specific technology under optimal conditions with a proprietary technology. Human uPAR Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # Q03405-1).
Catalog No. 50-253-4005 Supplier ACROBiosystems Supplier No. UPRHA2H925TESTS
APC-Labeled Human uPAR Protein His Tag (UPR-HA2H9) is produced via conjugation of APC to Human uPAR Protein His Tag with a new generation site-specific technology under optimal conditions with a proprietary technology. Human uPAR Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # Q03405-1).