Learn More
Biomatik Corporation Human Urate Transporter 1 (URAT1) ELISA Kit
Supplier:  Biomatik Corporation EKU08044
Encompass_Preferred
Human Urate Transporter 1 (URAT1) ELISA Kit for detecting URAT1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.127ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96S37. Gene ID: 116085. Target Synonyms: SLC22A12; OAT4L; RST; Solute Carrier Family 22 Member 12,Organic Anion/Urate Transporter; Organic anion transporter 4-like; Urate anion exchanger 1; Renal-specific transporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8701