Learn More
Biomatik Corporation Human Uridine Cytidine Kinase 2 (UCK2) ELISA Kit
Supplier:  Biomatik Corporation EKU11508
Encompass_Preferred
Human Uridine Cytidine Kinase 2 (UCK2) ELISA Kit for detecting UCK2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BZX2. Gene ID: 7371. Target Synonyms: UK; UMPK; Uridine Monophosphate Kinase; Cytidine monophosphokinase 2; Testis-specific protein TSA903; Uridine monophosphokinase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6746