Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Urinary TIM-1/KIM-1/HAVCR Quantikine ELISA Kit

Catalog No. p-7237475
Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
This item is not returnable. View return policy
This item is not returnable. View return policy

Human Urinary TIM-1/KIM-1/HAVCR Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Urinary TIM-1/KIM-1/HAVCR Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human TIM-1/KIM-1/HAVCR in urine (50 ul).

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.046 ng/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Urine
Specificity Natural and Recombinant Human TIM-1
For Use With (Application) ELISA
Gene ID (Entrez) 26762
Gene Symbol HAVCR1
Quantity 1 Kit
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1 TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Target TIM-1/KIM-1/HAVCR
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.