Learn More
Bioworld HUMAN URINARY TRYPSIN INHIB
Catalog No. 50253608
Encompass_Preferred
Human Urinary Trypsin Inhibitor (UTI) Source: Human Urine. Background: Urinary Trypsin Inhibitor (UTI, Ulinastatin) is a kind of glucoprotein derived from human urine. As a proteinase inhibitor, Ulinastatin inhibits the activity of t...
Catalog No. 50-253-608 Supplier Bioworld Supplier No. 220605102
Human Urinary Trypsin Inhibitor (UTI) Source: Human Urine. Background: Urinary Trypsin Inhibitor (UTI, Ulinastatin) is a kind of glucoprotein derived from human urine. As a proteinase inhibitor, Ulinastatin inhibits the activity of t...