Learn More
Biomatik Corporation Human Urocortin 3 (UCN3) ELISA Kit
Supplier:  Biomatik Corporation EKU08594
Encompass_Preferred
Human Urocortin 3 (UCN3) ELISA Kit for detecting UCN3 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 24.69-2000pg/ml. Sensitivity: 9.81pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q969E3. Gene ID: 114131. Target Synonyms: SCP; SPC; UCNIII; Stresscopin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4530