Learn More
Raybiotech Inc Human/Mouse/Rat Urocortin EIA Kit.
Supplier:  Raybiotech Inc EIAURO1
Encompass_Preferred
Optimized for serum. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-194-8849