Learn More
BIORTUS USA INC Human USP10 Protein, 100UG
Catalog No. NC2953437
Encompass_Preferred
Gene Name: USP10. Uniprot: Q14694. Construct: 8His-GST-TEV-GG-USP10(A2-L798 end). Expression Host: Insect cell. Purity: 80%. 1 each contains 100UG.
Catalog No. NC2953437 Supplier BIORTUS USA INC Supplier No. BP2801012A
Gene Name: USP10. Uniprot: Q14694. Construct: 8His-GST-TEV-GG-USP10(A2-L798 end). Expression Host: Insect cell. Purity: 80%. 1 each contains 100UG.