Learn More
BIORTUS USA INC Human USP48 Protein, 100UG
Catalog No. NC2953445
Encompass_Preferred
Gene Name: USP48. Uniprot: Q86UV5. Construct: USP48(A2-H1035 end)-Flag. Expression Host: Insect cell. Purity: 85%.
Catalog No. NC2953445 Supplier BIORTUS USA INC Supplier No. BP1244304A
Gene Name: USP48. Uniprot: Q86UV5. Construct: USP48(A2-H1035 end)-Flag. Expression Host: Insect cell. Purity: 85%.