Learn More
Biomatik Corporation Human USP6 N-Terminal Like Protein (USP6NL) ELISA Kit
Supplier:  Biomatik Corporation EKU08065
Encompass_Preferred
Human USP6 N-Terminal Like Protein (USP6NL) ELISA Kit for detecting USP6NL in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92738. Gene ID: 9712. Target Synonyms: RNTRE; TRE2NL; Related to the N-terminus of tre. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6110