Learn More
BIOHIPPO Human USP6NL Microsample
SDP
Catalog No. 5000314747
Encompass
Human USP6NL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of USP6NL in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human USP6NL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human USP6NL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human USP6NL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14747 Supplier BIOHIPPO Supplier No. BHE15211115
Human USP6NL Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of USP6NL in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human USP6NL Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human USP6NL Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human USP6NL biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color