Learn More
BIORTUS USA INC Human USP7 Protein, 100UG
Catalog No. NC2953447
Encompass_Preferred
Gene Name: USP7. Uniprot: Q93009. Construct:His-TEV-USP7(K208-E560). Expression Host: E. coli. Purity: 85%.
Catalog No. NC2953447 Supplier BIORTUS USA INC Supplier No. BP0599001A
Gene Name: USP7. Uniprot: Q93009. Construct:His-TEV-USP7(K208-E560). Expression Host: E. coli. Purity: 85%.