Learn More
Sino Biological Human USP7 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11681MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human USP7 / HAUSP (rh USP7 / HAUSP; Catalog#11681-H20B; NP_003461.2; Lys208-Glu560). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3053