Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Uteroglobin Recombinant Protein, PeproTech®

Catalog No. 1501850UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
150-18-50UG 50 μg
15018100UG 100 μg
150-18-10UG 10 μg
150-18-1MG 1 mg
15018250UG 250 μg
15018500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 150-18-50UG Supplier Gibco™ Supplier No. 1501850UG
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Uteroglobin is a 16.1 kDa homodimeric protein consisting of 142 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Uteroglobin, which is a member of the Secretoglobin superfamily and is also known as Clara cell phospholipid-binding protein, is a multifunctional protein that can exert anti-inflammatory and anti-tumorigenic effects by binding small hydrophobic molecules such as phospholipids and prostaglandins. The small, non-glycosylated protein named for its high levels of expression in pre-implantation embryos, where it exhibits growth stimulatory effects, is produced and secreted by the non-ciliated, non-mucous Clara cells predominant in the epithelial surfaces of pulmonary airways, as well as other non-ciliated epithelia. Members of the Secretoglobin superfamily demonstrate a high level of structural conservation and are characterized as small, secretory homo- or heterodimers. In addition to sequestering pro-inflammatory mediators and carcinogens, Uteroglobin has been implicated in the inhibition of cell migration and invasion, platelet aggregation, and T cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11684
For Use With (Application) Control, Western Blot Control, Western Blot
Formulation Protein with no preservative
Gene ID (Entrez) 7356
Molecular Weight (g/mol) 16.1 kDa
Name Human Uteroglobin
Purification Method Purified
Quantity 50 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias Blastokinin; CC10; CC16; CCPBP; CCSP; Clara cell 17 kDa protein; clara cell phospholipid-binding protein; clara cell secretory protein; clara cells 10 kDa secretory protein; PCBB; PCB-binding protein; PCB-BP; SCGB1A1; Secretoglobin family 1A member 1; secretoglobin, family 1A, member 1 (uteroglobin); UG; Ugb; UP1; UP-1; Urinary protein 1; urine protein 1; uteroglobin; Uteroglobin (Clara cell secretory protein); Utg
Common Name Uteroglobin
Gene Symbol SCGB1A1
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MEICPSFQRV IETLLMDTPS SYEAAMELFS PDQDMREAGA QLKKLVDTLP QKPRESIIKL MEKIAQSSLC N
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.