Learn More
Biomatik Corporation Human Vaccinia Related Kinase 2 (VRK2) ELISA Kit
Supplier:  Biomatik Corporation EKU12408
Encompass_Preferred
Human Vaccinia Related Kinase 2 (VRK2) ELISA Kit for detecting VRK2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86Y07. Gene ID: 7444. Target Synonyms: Serine/threonine-protein kinase VRK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5962