Learn More
Biomatik Corporation Human Valosin Containing Protein (VCP) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12001
Encompass_Preferred
Human Valosin Containing Protein (VCP) Mini sample ELISA Kit for detecting VCP in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.127ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55072. Target Synonyms: IBMPFD; p97; TERA; Transitional Endoplasmic Reticulum ATPase; 15S Mg (2+)-ATPase p97 subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6198