Learn More
Creative Biomart Human Varicella Zoster Immune Globulin
Catalog No. 502084228
Encompass_Preferred
The product is a solvent/detergent-treated sterile liquid preparation of purified human immune globulin G (IgG) containing antibodies to varicella zoster virus (anti-VZV). It is prepared from pools of healthy human donors' plasma via anion-exchange column chromatography.
Catalog No. 50-208-4228 Supplier Creative Biomart Supplier No. THP0177
The product is a solvent/detergent-treated sterile liquid preparation of purified human immune globulin G (IgG) containing antibodies to varicella zoster virus (anti-VZV). It is prepared from pools of healthy human donors' plasma via anion-exchange column chromatography.