Learn More
Biomatik Corporation Human Vascular Cell Adhesion Molecule 1 (VCAM1) CLIA Kit
Supplier:  Biomatik Corporation EKU09467
Encompass_Preferred
Human Vascular Cell Adhesion Molecule 1 (VCAM1) CLIA Kit for detecting VCAM1 in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 68.6-50,000pg/ml. Sensitivity: 27.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P19320. Gene ID: 7412. Target Synonyms: CD106; INCAM-100; L1CAM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5963