Learn More
American Research Products Inc Human Vascular endothelial cel
Catalog No. 5000329123
Encompass
Human Vascular endothelial cell growth factor receptor 2 VEGFR-2/Flk-1 ELISA kit from CUSABIO Detection Range 0 39 ng/mL-25 ng/mL Sensitivity 0 098 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29123 Supplier American Research Products Inc Supplier No. CSBE04763H
Human Vascular endothelial cell growth factor receptor 2 VEGFR-2/Flk-1 ELISA kit from CUSABIO Detection Range 0 39 ng/mL-25 ng/mL Sensitivity 0 098 ng/mL Sample serum plasma tissue homogenates Method Sandwich