Learn More
Biomatik Corporation Human Vascular Endothelial Growth Factor C (VEGFC) CLIA Kit
Supplier:  Biomatik Corporation EKU08851
Encompass_Preferred
Human Vascular Endothelial Growth Factor C (VEGFC) CLIA Kit for detecting VEGFC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.92pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P49767. Gene ID: 7424. Target Synonyms: VEGF-C; Flt4-L; VRPL; Vascular Endothelial Growth Factor-Related Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5477