Learn More
Biomatik Corporation Human Vasodilator Stimulated Phosphoprotein (VASP) ELISA Kit
Supplier:  Biomatik Corporation EKU08117
Encompass_Preferred
Human Vasodilator Stimulated Phosphoprotein (VASP) ELISA Kit for detecting VASP in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50552. Gene ID: 7408. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8697