Learn More
Invitrogen™ Human Vasohibin-1 ELISA Kit
Description
Human Vasohibin-1 quantitates human Vasohibin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Vasohibin-1.
Specifications
Specifications
| Accession Number | AI834978; D930046M13Rik; G630009D10Rik; KIAA1036; RGD1564082; TTCP 1; Tubulin carboxypeptidase 1; Tubulinyl-Tyr carboxypeptidase 1; Tyrosine carboxypeptidase 1; VASH; VASH1; vasohibin 1; vasohibin-1 |
| Assay Range | 11 to 2,500 pg/mL |
| Assay Sensitivity | 11 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 22846 |
| Gene Symbol | VASH1 |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.