Learn More
ACROBiosystems ActiveMax Human VCAM-1 / CD106 μBeads, premium grade (for cells)
Catalog No. 502719178
Encompass_Preferred
ActiveMax Human VCAM-1 / CD106 μBeads, premium grade (for cells) are produced under sterile manufacturing conditions (ISO 5) , and no animal- or human-derived components are used throughout the production process. It is produced under our rigorous quality control system that includes a comprehensive set of tests including sterility and endotoxin tests.
Catalog No. 50-271-9178 Supplier ACROBiosystems Supplier No. MBSC01910MG
Add to Cart
Add to Cart
Request a Quote
ActiveMax Human VCAM-1 / CD106 μBeads, premium grade (for cells) are produced under sterile manufacturing conditions (ISO 5) , and no animal- or human-derived components are used throughout the production process. It is produced under our rigorous quality control system that includes a comprehensive set of tests including sterility and endotoxin tests.