Learn More
Invitrogen™ Human VCAM-1 Recombinant Protein
Description
RP-75753 contains purified recombinant human VCAM-1 protein with an N-terminal hexahistidine tag. RP-75753 may be used as a positive control in many immunological assays.
Specifications
Specifications
| Accession Number | P19320 |
| For Use With (Application) | Control, Western Blot |
| Formulation | 20 mM tris HCl with 50% glycerol, 5 mM EDTA and no preservative; pH 8 |
| Gene ID (Entrez) | 7412 |
| Name | Human VCAM-1 |
| Quantity | 1 mg |
| Source | E. coli |
| Immunogen | N/A |
| Regulatory Status | RUO |
| Gene Alias | adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.