Learn More
Sino Biological Anti-VCAM-1/CD106 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10113T08100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human VCAM-1/CD106 (rh VCAM-1/CD106; Catalog#10113-H07H; NP_001069.1; Phe25-Pro697). VCAM-1/CD106 specific IgG was purified by Human VCAM-1/CD106 affinity chromatography.
Catalog No. 50-255-5468