Learn More
Sino Biological Recombinant Human VCAM-1/CD106 Protein (hFc Tag), Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 10113H02HB20
Encompass_Preferred
A DNA sequence encoding the human VCAM1 (NP_001069.1) (Met 1-Pro 697) was expressed with the Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5469